Search
Search
| Gene Symbol: CCDC34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003122907.4 | XP_003122955.1 | 444 | 80 | ||
| GenBank mRNA | AK234803.1 | - | 1 - 2 | 392 | 80 | |
| AK347457.1 | - | 1 - 2 | 363 | 80 | ||
Set Membership: |
|