Search
Search
| Gene Symbol: NMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021066846.1 | XP_020922505.1 | 524 | 66 | ||
| GenBank mRNA | AK235665.1 | - | 4 - 5 | 510 | 66 | |
| AK389663.1 | - | 4 - 5 | 518 | 66 | ||
Set Membership: |
|