Search
Search

SERPINB7| Gene Symbol: SERPINB7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001925047.5 | XP_001925082.1 | 687 | 102 | ||
| GenBank mRNA | AK230691.1 | - | 3 - 4 | 276 | 102 | |
| AK231651.1 | - | 1 - 2 | 196 | 102 | ||
Set Membership: |
|