Search
Search
| Gene Symbol: PRRC2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003130303.6 | XP_003130351.3 | 2619 | 84 | ||
| XM_005653732.2 | XP_005653789.1 | 2624 | 84 | |||
| XM_005653733.3 | XP_005653790.1 | 2608 | 84 | |||
| XM_005653735.2 | XP_005653792.1 | 2619 | 84 | |||
| XM_005653736.3 | XP_005653793.1 | 2608 | 84 | |||
| XM_005653737.3 | XP_005653794.1 | 2583 | 84 | |||
| XM_005653738.2 | XP_005653795.1 | 2619 | 84 | |||
| XM_005653739.3 | XP_005653796.1 | 2608 | 84 | |||
| XM_005653740.3 | XP_005653797.1 | 2608 | 84 | |||
| XM_005653741.3 | XP_005653798.1 | 2608 | 84 | |||
| XM_005653742.3 | XP_005653799.1 | 2608 | 84 | |||
| XM_013977753.2 | XP_013833207.1 | 2577 | 84 | |||
| XM_013977760.2 | XP_013833214.1 | 2608 | 84 | |||
| XM_021063628.1 | XP_020919287.1 | 2608 | 84 | |||
| XM_021063629.1 | XP_020919288.1 | 2608 | 84 | |||
| XM_021063630.1 | XP_020919289.1 | 2608 | 84 | |||
| XM_021063631.1 | XP_020919290.1 | 2608 | 84 | |||
| XM_021063632.1 | XP_020919291.1 | 2608 | 84 | |||
| XM_021063633.1 | XP_020919292.1 | 2608 | 84 | |||
| XM_021063634.1 | XP_020919293.1 | 2608 | 84 | |||
| GenBank mRNA | AK230791.1 | - | 3 - 4 | 663 | 84 | |
| AK393649.1 | - | 3 - 4 | 760 | 84 | ||
Set Membership: |
|