Search
Search

LOC100738955| Gene Symbol: LOC100738955 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021095981.1 | XP_020951640.1 | 117 | |||
| GenBank mRNA | AK236082.1 | - | 4 - 5 | 484 | 117 | |
| AK400165.1 | - | 4 - 5 | 117 | |||
Set Membership: |
|