Search
Search
| Gene Symbol: IAH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003481289.4 | XP_003481337.1 | 196 | 132 | ||
| GenBank mRNA | AK236643.1 | - | 2 - 3 | 186 | 132 | |
| AK350224.1 | - | 2 - 3 | 166 | 132 | ||
Set Membership: |
|