Search
Search
| Gene Symbol: USP32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021067305.1 | XP_020922964.1 | 1630 | 59 | ||
| XM_021067306.1 | XP_020922965.1 | 1516 | 59 | |||
| GenBank mRNA | AK236959.1 | - | 13 - 14 | 1620 | 59 | |
| AK345961.1 | - | 13 - 14 | 1622 | 59 | ||
Set Membership: |
|