Search
Search
| Gene Symbol: SETX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013987041.2 | XP_013842495.1 | 751 | 64 | ||
| XM_013987048.2 | XP_013842502.1 | 755 | 64 | |||
| XM_021070644.1 | XP_020926303.1 | 647 | 64 | |||
| XM_021070651.1 | XP_020926310.1 | 651 | 64 | |||
| XM_021070654.1 | XP_020926313.1 | 752 | 64 | |||
| XM_021070661.1 | XP_020926320.1 | 758 | 64 | |||
| XM_021070670.1 | XP_020926329.1 | 751 | 64 | |||
| XM_021070681.1 | XP_020926340.1 | 751 | 64 | |||
| XM_021070690.1 | XP_020926349.1 | 752 | 64 | |||
| XM_021070705.1 | XP_020926364.1 | 751 | 64 | |||
| XM_021070714.1 | XP_020926373.1 | 751 | 64 | |||
| XM_021070725.1 | XP_020926384.1 | 755 | 64 | |||
| XM_021070731.1 | XP_020926390.1 | 759 | 64 | |||
| GenBank mRNA | AK231150.1 | - | 5 - 6 | 682 | 64 | |
Set Membership: |
|