Search
Search
| Gene Symbol: EIF2S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928339.4 | XP_001928374.1 | 415 | 152 | ||
| GenBank mRNA | AK239598.1 | - | 1 - 2 | 134 | 152 | |
| AK351968.1 | - | 1 - 2 | 121 | 152 | ||
| AK391511.1 | - | 1 - 2 | 131 | 152 | ||
| AK394680.1 | - | 1 - 2 | 136 | 152 | ||
| AY609835.1 | - | 1 - 2 | 107 | 152 | ||
Set Membership: |
|