Search
Search
| Gene Symbol: POLR3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021089946.1 | XP_020945605.1 | 916 | 80 | ||
| GenBank mRNA | AK239631.1 | - | 1 - 2 | 479 | 80 | |
| AK390468.1 | - | 1 - 2 | 486 | 80 | ||
Set Membership: |
|