Search
Search
| Gene Symbol: TGFBI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021084784.1 | XP_020940443.1 | 2058 | 105 | ||
| GenBank mRNA | AK240211.1 | - | 15 - 16 | 2043 | 105 | |
| AK399297.1 | - | 10 - 11 | 1347 | 105 | ||
Set Membership: |
|