Search
Search
| Gene Symbol: MYNN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003358669.4 | XP_003358717.1 | 2175 | 120 | ||
| GenBank mRNA | AK343837.1 | - | 6 - 7 | 1568 | 120 | |
| KY470829.1 | - | 5 - 6 | 1514 | 120 | ||
Set Membership: |
|