Search
Search
| Gene Symbol: CEP41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003134674.6 | XP_003134722.3 | 579 | 100 | ||
| XM_021078823.1 | XP_020934482.1 | 636 | 100 | |||
| GenBank mRNA | AK347647.1 | - | 6 - 7 | 483 | 100 | |
Set Membership: |
|