Search
Search
| Gene Symbol: STRADB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003133587.5 | XP_003133635.2 | 447 | 67 | ||
| XM_005654191.3 | XP_005654248.1 | 405 | 67 | |||
| XM_005654192.3 | XP_005654249.1 | 418 | 67 | |||
| XM_013982348.2 | XP_013837802.1 | 276 | 67 | |||
| XM_013982357.2 | XP_013837811.1 | 418 | 67 | |||
| XM_021075700.1 | XP_020931359.1 | 222 | 67 | |||
| XM_021075701.1 | XP_020931360.1 | 332 | 67 | |||
| GenBank mRNA | AK348529.1 | - | 2 - 3 | 337 | 67 | |
| AK348997.1 | - | 3 - 4 | 329 | 67 | ||
| AK397647.1 | - | 3 - 4 | 381 | 67 | ||
| AY609457.1 | - | 3 - 4 | 351 | 67 | ||
Set Membership: |
|