Search
Search
| Gene Symbol: ADGRL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021083697.1 | XP_020939356.1 | 7973 | 106 | ||
| XM_021083699.1 | XP_020939358.1 | 7650 | 106 | |||
| XM_021083700.1 | XP_020939359.1 | 7831 | 106 | |||
| GenBank mRNA | AK389348.1 | - | 1 | 2014 | 106 | |
Set Membership: |
|