Search
Search
| Gene Symbol: SNX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021090785.1 | XP_020946444.1 | 926 | 124 | ||
| XM_021090786.1 | XP_020946445.1 | 860 | 124 | |||
| GenBank mRNA | AK231595.1 | - | 2 - 3 | 526 | 124 | |
| AK238761.1 | - | 2 - 3 | 360 | 124 | ||
| AK390011.1 | - | 2 - 3 | 559 | 124 | ||
| AK397053.1 | - | 2 - 3 | 574 | 124 | ||
Set Membership: |
|