Search
Search
| Gene Symbol: ABHD11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005661982.3 | XP_005662039.1 | 374 | 87 | ||
| XM_013995544.1 | XP_013850998.1 | 338 | 87 | |||
| GenBank mRNA | AK231855.1 | - | 2 - 3 | 315 | 87 | |
| AK347674.1 | - | 2 - 3 | 320 | 87 | ||
| AK394477.1 | - | 2 - 3 | 315 | 87 | ||
| AK397126.1 | - | 2 - 3 | 343 | 87 | ||
Set Membership: |
|