Search
Search

RETREG1| Gene Symbol: RETREG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003359728.5 | XP_003359776.2 | 1276 | 81 | ||
| XM_003483804.4 | XP_003483852.1 | 1030 | 81 | |||
| XM_005672396.3 | XP_005672453.1 | 729 | 81 | |||
| GenBank mRNA | AK231960.1 | - | 7 - 8 | 916 | 81 | |
| AK232203.1 | - | 5 - 6 | 1039 | 81 | ||
| AK390068.1 | - | 7 - 8 | 1035 | 81 | ||
| AK395660.1 | - | 5 - 6 | 1040 | 81 | ||
| AK400545.1 | - | 7 - 8 | 993 | 81 | ||
| AK400671.1 | - | 6 - 7 | 729 | 81 | ||
| JX854456.1 | - | 7 - 8 | 672 | 81 | ||
Set Membership: |
|