Search
Search
| Gene Symbol: BACE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005670363.3 | XP_005670420.1 | 530 | 83 | ||
| XM_021071002.1 | XP_020926661.1 | 530 | 83 | |||
| XM_021071004.1 | XP_020926663.1 | 530 | 83 | |||
| XM_021071005.1 | XP_020926664.1 | 530 | 83 | |||
| XM_021071006.1 | XP_020926665.1 | 530 | 83 | |||
| GenBank mRNA | AK232199.1 | - | 2 - 3 | 535 | 83 | |
Set Membership: |
|