Search
Search
| Gene Symbol: CEMIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021064787.1 | XP_020920446.1 | 4457 | 98 | ||
| XM_021064792.1 | XP_020920451.1 | 3363 | 98 | |||
| XM_021064799.1 | XP_020920458.1 | 3236 | 98 | |||
| XM_021064806.1 | XP_020920465.1 | 3783 | 98 | |||
| GenBank mRNA | AK389661.1 | - | 5 - 6 | 802 | 98 | |
Set Membership: |
|