Search
Search
| Gene Symbol: EML4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003125196.6 | XP_003125244.4 | 4357 | 78 | ||
| XM_005662587.3 | XP_005662644.1 | 4960 | 78 | |||
| XM_005662588.3 | XP_005662645.1 | 4921 | 78 | |||
| XM_013996185.2 | XP_013851639.1 | 5054 | 78 | |||
| XM_013996186.2 | XP_013851640.1 | 5017 | 78 | |||
| XM_021087581.1 | XP_020943240.1 | 4325 | 78 | |||
| XM_021087582.1 | XP_020943241.1 | 4150 | 78 | |||
| GenBank mRNA | AK389880.1 | - | 1 | 120 | 78 | |
Set Membership: |
|