Search
Search
| Gene Symbol: GIT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021067515.1 | XP_020923174.1 | 2165 | 66 | ||
| XM_021067516.1 | XP_020923175.1 | 2157 | 66 | |||
| GenBank mRNA | AK390481.1 | - | 5 - 6 | 598 | 66 | |
Set Membership: |
|