Search
Search
| Gene Symbol: ELMOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013979345.2 | XP_013834799.2 | 398 | 109 | ||
| XM_021062732.1 | XP_020918391.1 | 314 | 109 | |||
| XM_021062733.1 | XP_020918392.1 | 1391 | 109 | |||
| XM_021062734.1 | XP_020918393.1 | 465 | 109 | |||
| XM_021062735.1 | XP_020918394.1 | 103 | 109 | |||
| GenBank mRNA | AK393336.1 | - | 2 - 3 | 279 | 109 | |
Set Membership: |
|