Search
Search

PHACTR1| Gene Symbol: PHACTR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021099759.1 | XP_020955418.1 | 1932 | 89 | ||
| XM_021099765.1 | XP_020955424.1 | 1930 | 89 | |||
| GenBank mRNA | AK393890.1 | - | 1 - 2 | 111 | 89 | |
Set Membership: |
|