Search
Search
| Gene Symbol: ETFDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003129003.4 | XP_003129051.1 | 1899 | 113 | ||
| XM_005666668.3 | XP_005666725.1 | 1631 | 113 | |||
| XM_005666669.3 | XP_005666726.1 | 1524 | 113 | |||
| GenBank mRNA | AK394624.1 | - | 7 - 8 | 898 | 113 | |
Set Membership: |
|