Search
Search
| Gene Symbol: QSER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013994460.2 | XP_013849914.1 | 5732 | 108 | ||
| XM_021083210.1 | XP_020938869.1 | 5927 | 108 | |||
| XM_021083211.1 | XP_020938870.1 | 8569 | 108 | |||
| XM_021083212.1 | XP_020938871.1 | 4602 | 108 | |||
| XM_021083213.1 | XP_020938872.1 | 5862 | 108 | |||
| XM_021083215.1 | XP_020938874.1 | 8407 | 108 | |||
| GenBank mRNA | AK394704.1 | - | 4 - 5 | 632 | 108 | |
Set Membership: |
|