Search
Search
| Gene Symbol: COBLL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021075409.1 | XP_020931068.1 | 1688 | 102 | ||
| XM_021075410.1 | XP_020931069.1 | 1688 | 102 | |||
| XM_021075411.1 | XP_020931070.1 | 1688 | 102 | |||
| XM_021075412.1 | XP_020931071.1 | 1527 | 102 | |||
| XM_021075413.1 | XP_020931072.1 | 1546 | 102 | |||
| XM_021075414.1 | XP_020931073.1 | 1716 | 102 | |||
| XM_021075415.1 | XP_020931074.1 | 1499 | 102 | |||
| XM_021075416.1 | XP_020931075.1 | 1444 | 102 | |||
| XM_021075417.1 | XP_020931076.1 | 1283 | 102 | |||
| XM_021075418.1 | XP_020931077.1 | 1283 | 102 | |||
| XM_021075419.1 | XP_020931078.1 | 1358 | 102 | |||
| XM_021075420.1 | XP_020931079.1 | 2550 | 102 | |||
| XM_021075421.1 | XP_020931080.1 | 1394 | 102 | |||
| XM_021075422.1 | XP_020931081.1 | 1194 | 102 | |||
| XM_021075423.1 | XP_020931082.1 | 4479 | 102 | |||
| XM_021075424.1 | XP_020931083.1 | 1234 | 102 | |||
| XM_021075425.1 | XP_020931084.1 | 3029 | 102 | |||
| GenBank mRNA | AK395427.1 | - | 8 - 9 | 1242 | 102 | |
Set Membership: |
|