Search
Search
| Gene Symbol: TRIM33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021090006.1 | XP_020945665.1 | 632 | 92 | ||
| XM_021090007.1 | XP_020945666.1 | 521 | 92 | |||
| XM_021090008.1 | XP_020945667.1 | 632 | 92 | |||
| GenBank mRNA | AK395664.1 | - | 1 - 2 | 638 | 92 | |
Set Membership: |
|