Search
Search
| Gene Symbol: BRD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021094213.1 | XP_020949872.1 | 682 | 103 | ||
| XM_021094214.1 | XP_020949873.1 | 687 | 103 | |||
| GenBank mRNA | AK395749.1 | - | 4 - 5 | 687 | 103 | |
Set Membership: |
|