Search
Search
| Gene Symbol: GALNT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021081725.1 | XP_020937384.1 | 1514 | 109 | ||
| XM_021081726.1 | XP_020937385.1 | 1610 | 109 | |||
| XM_021081727.1 | XP_020937386.1 | 1499 | 109 | |||
| GenBank mRNA | AK397516.1 | - | 50 | 109 | ||
Set Membership: |
|