Search
Search
| Gene Symbol: SORL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021063009.1 | XP_020918668.1 | 5057 | 69 | ||
| XM_021063010.1 | XP_020918669.1 | 5056 | 69 | |||
| XM_021063011.1 | XP_020918670.1 | 5148 | 69 | |||
| XM_021063012.1 | XP_020918671.1 | 5126 | 69 | |||
| XM_021063013.1 | XP_020918672.1 | 5126 | 69 | |||
| XM_021063014.1 | XP_020918673.1 | 4920 | 69 | |||
| XM_021063015.1 | XP_020918674.1 | 5158 | 69 | |||
| GenBank mRNA | AK397605.1 | - | 6 - 7 | 761 | 69 | |
Set Membership: |
|