Search
Search
| Gene Symbol: PHYHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928841.5 | XP_001928876.1 | 554 | 77 | ||
| XM_013993922.2 | XP_013849376.1 | 1215 | 77 | |||
| GenBank mRNA | AK232373.1 | - | 5 - 6 | 454 | 77 | |
| AK236678.1 | - | 5 - 6 | 449 | 77 | ||
| AK346009.1 | - | 5 - 6 | 447 | 77 | ||
Set Membership: |
|