Search
Search

DPY19L4| Gene Symbol: DPY19L4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001926300.6 | XP_001926335.1 | 1230 | 140 | ||
| XM_005662961.3 | XP_005663018.1 | 1101 | 140 | |||
| XM_021089085.1 | XP_020944744.1 | 1297 | 140 | |||
| XM_021089086.1 | XP_020944745.1 | 1016 | 140 | |||
Set Membership: |
|