Search
Search
| Gene Symbol: CNOT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005664822.3 | XP_005664879.1 | 4050 | 85 | ||
| XM_013998750.2 | XP_013854204.1 | 4050 | 85 | |||
| XM_013998751.2 | XP_013854205.1 | 3497 | 85 | |||
| XM_013998752.2 | XP_013854206.1 | 4047 | 85 | |||
| XM_013998753.2 | XP_013854207.1 | 4047 | 85 | |||
| XM_013998754.2 | XP_013854208.1 | 3921 | 85 | |||
| XM_021094881.1 | XP_020950540.1 | 5485 | 85 | |||
| XM_021094882.1 | XP_020950541.1 | 3918 | 85 | |||
Set Membership: |
|