Search
Search
| Gene Symbol: HEPH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005673694.3 | XP_005673751.1 | 1087 | 69 | ||
| XM_013986223.2 | XP_013841677.1 | 2388 | 69 | |||
| XM_013986224.2 | XP_013841678.1 | 2391 | 69 | |||
| XM_021079773.1 | XP_020935432.1 | 1092 | 69 | |||
| XM_021079774.1 | XP_020935433.1 | 1089 | 69 | |||
| XM_021079775.1 | XP_020935434.1 | 1093 | 69 | |||
| XM_021079776.1 | XP_020935435.1 | 1090 | 69 | |||
Set Membership: |
|