Search
Search
| Gene Symbol: GRAMD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021091362.1 | XP_020947021.1 | 880 | 99 | ||
| XM_021091363.1 | XP_020947022.1 | 780 | 99 | |||
| XM_021091364.1 | XP_020947023.1 | 694 | 99 | |||
| XM_021091365.1 | XP_020947024.1 | 787 | 99 | |||
| XM_021091366.1 | XP_020947025.1 | 784 | 99 | |||
| XM_021091367.1 | XP_020947026.1 | 700 | 99 | |||
| XM_021091368.1 | XP_020947027.1 | 733 | 99 | |||
| XM_021091369.1 | XP_020947028.1 | 480 | 99 | |||