Search
Search
| Gene Symbol: TULP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003126566.4 | XP_003126614.1 | 979 | 162 | ||
| XM_005664076.3 | XP_005664133.1 | 978 | 162 | |||
| XM_013997835.2 | XP_013853289.1 | 971 | 162 | |||
Set Membership: |
|