Search
Search

poc1b| Gene Symbol: poc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004988.1 | NP_001004988.1 | 5 - 6 | 649 | 92 | |
| XM_012958840.2 | XP_012814294.1 | 5 - 6 | 767 | 92 | ||
| XM_012958841.2 | XP_012814295.1 | 5 - 6 | 769 | 92 | ||
| XM_012958842.2 | XP_012814296.1 | 5 - 6 | 766 | 92 | ||
| XM_012958844.1 | XP_012814298.1 | 5 - 6 | 621 | 92 | ||
| XM_018092031.1 | XP_017947520.1 | 4 - 5 | 544 | 92 | ||
| XM_018092032.1 | XP_017947521.1 | 5 - 6 | 766 | 92 | ||
| GenBank mRNA | BC075548.1 | - | 5 - 6 | 649 | 92 | |
| CU024965.1 | - | 5 - 6 | 700 | 92 | ||
Set Membership: |
|