Search
Search

spc24| Gene Symbol: spc24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203726.1 | NP_989057.1 | 3 - 4 | 460 | 95 | |
| GenBank mRNA | BC061328.1 | - | 3 - 4 | 460 | 95 | |
| CR761127.2 | - | 2 - 3 | 391 | 95 | ||
Set Membership: |
|