Search
Search

msrb2| Gene Symbol: msrb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015905.2 | NP_001015905.1 | 4 - 5 | 547 | 64 | |
| GenBank mRNA | BC121263.1 | - | 3 - 4 | 239 | 64 | |
| BC170741.1 | - | 4 - 5 | 528 | 64 | ||
| BC171252.1 | - | 4 - 5 | 528 | 64 | ||
| CR926437.2 | - | 4 - 5 | 547 | 64 | ||
Set Membership: |
|