Search
Search

pigu| Gene Symbol: pigu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016206.2 | NP_001016206.1 | 1 - 2 | 186 | 61 | |
| GenBank mRNA | BC170796.1 | - | 1 - 2 | 156 | 61 | |
| BC170798.1 | - | 1 - 2 | 156 | 61 | ||
| CR926339.2 | - | 1 - 2 | 186 | 61 | ||
Set Membership: |
|