Search
Search

crtap| Gene Symbol: crtap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016647.2 | NP_001016647.1 | 2 - 3 | 996 | 78 | |
| GenBank mRNA | BC135296.1 | - | 2 - 3 | 1050 | 78 | |
| CR760380.2 | - | 2 - 3 | 996 | 78 | ||
Set Membership: |
|