Search
Search

slc18a1| Gene Symbol: slc18a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012959124.2 | XP_012814578.1 | 10 - 11 | 1128 | 82 | |
| XM_018092118.1 | XP_017947607.1 | 9 - 10 | 1161 | 82 | ||
| GenBank mRNA | BC090568.1 | - | 9 - 10 | 967 | 82 | |
| CT025350.1 | - | 9 - 10 | 980 | 82 | ||
Set Membership: |
|