Search
Search

hdac1| Gene Symbol: hdac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030393.1 | NP_001025564.1 | 1 - 2 | 74 | 82 | |
| GenBank mRNA | BC090604.1 | - | 1 - 2 | 74 | 82 | |
| CT030593.1 | - | 1 - 2 | 52 | 82 | ||
Set Membership: |
|