Search
Search

atp2a1| Gene Symbol: atp2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032327.1 | NP_001027498.1 | 12 - 13 | 1595 | 85 | |
| XM_012954931.2 | XP_012810385.1 | 12 - 13 | 1627 | 85 | ||
| GenBank mRNA | BC096519.1 | - | 12 - 13 | 1595 | 85 | |
Set Membership: |
|