Search
Search

mau2| Gene Symbol: mau2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015927.2 | NP_001015927.1 | 2 - 3 | 272 | 70 | |
| XM_012961796.2 | XP_012817250.1 | 2 - 3 | 316 | 70 | ||
| GenBank mRNA | BC160796.1 | - | 2 - 3 | 290 | 70 | |
| CR926381.2 | - | 2 - 3 | 272 | 70 | ||
Set Membership: |
|