Search
Search

igfbp5| Gene Symbol: igfbp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016042.2 | NP_001016042.1 | 3 - 4 | 512 | 106 | |
| XM_012970400.2 | XP_012825854.1 | 4 - 5 | 1376 | 106 | ||
| GenBank mRNA | BC121208.1 | - | 3 - 4 | 1233 | 106 | |
| BC158970.1 | - | 4 - 5 | 1095 | 106 | ||
| BC161338.1 | - | 3 - 4 | 1442 | 106 | ||
| BC170988.1 | - | 3 - 4 | 680 | 106 | ||
| CR848090.2 | - | 3 - 4 | 512 | 106 | ||
Set Membership: |
|