Search
Search

paqr3| Gene Symbol: paqr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012962038.2 | XP_012817492.1 | 6 - 7 | 1031 | 78 | |
| XM_012962044.2 | XP_012817498.1 | 6 - 7 | 905 | 78 | ||
| XM_012962048.2 | XP_012817502.1 | 5 - 6 | 988 | 78 | ||
| GenBank mRNA | BC171300.1 | - | 5 - 6 | 792 | 78 | |
| CR762035.2 | - | 5 - 6 | 793 | 78 | ||
Set Membership: |
|