Search
Search

b4galt6| Gene Symbol: b4galt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016385.2 | NP_001016385.1 | 3 - 4 | 721 | 73 | |
| GenBank mRNA | BC170760.1 | - | 3 - 4 | 507 | 73 | |
| BC170764.1 | - | 3 - 4 | 507 | 73 | ||
| CR761207.2 | - | 3 - 4 | 721 | 73 | ||
Set Membership: |
|