Search
Search

mrpl53| Gene Symbol: mrpl53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005117.2 | NP_001005117.1 | 2 - 3 | 277 | 70 | |
| GenBank mRNA | BC077056.1 | - | 2 - 3 | 231 | 70 | |
| CR926208.2 | - | 2 - 3 | 277 | 70 | ||
Set Membership: |
|